Mouse Recombinant DNER extracellular domain (from HEK293 Cells)
Supplier: 0000027161
AG-40A-0177-C010 AG-40A-0177-C050
102515-330EA 263.08 USD
102515-330 102515-332
Mouse Recombinant DNER extracellular domain (from HEK293 Cells)
Proteins and Peptides
DNER is an activator of the Notch1 pathway. It mediates neuron-glia interaction during astrocytogenesis.