Anti-beta-Tubulin Human Antibody [clone: S11B]

Supplier: Adipogen
AG-27B-0008-C100
102514-704EA 415.38 USD
102514-704
Anti-beta-Tubulin Human Antibody [clone: S11B]
Antibodies
anti-beta-Tubulin, monoclonal antibody (recombinant)(S11B) is an antibody developed by antibody phage display technology using a human naive antibody gene library. These libraries consist of scFv (single chain fragment variable) composed of VH (variable domain of the human immunoglobulin heavy chain) and VL (variable domain of the human immunoglobulin light chain) connected by a polypeptide linker. The antibody fragments are displayed on the surface of filamentous bacteriophage (M13). This scFv was selected by affinity selection on antigen in a process termed panning. Multiple rounds of panning are performed to enrich for antigen-specific scFv-phage. Monoclonal antibodies are subsequently identified by screening after each round of selection. The selected monoclonal scFv is cloned into an appropriate vector containing a Fc portion of interest and then produced in mammalian cells to generate an IgG like scFv-Fc fusion protein._x000D_ _x000D_

  • Stable for at least 1 year after receipt when stored at -20°C.

Purified from HEK 293 cell culture supernatant.

Purity: >95% (SDS-PAGE)

Type:
Antigen: beta-Tubulin
Clonality:
Clone: S11B
Conjugation:
Epitope:
Host: Human
Isotype: IgG2 lambda
Reactivity: Drosophila, Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR