Anti-Giantin Human Antibody [clone: TA10]

Supplier: Adipogen
AG-27B-0003-C100
102514-694EA 415.38 USD
102514-694
Anti-Giantin Human Antibody [clone: TA10]
Antibodies
Giantin, the largest Golgi membrane protein, is C-terminally anchored to cis- and medial-Golgi membranes and forms a long ~250nm coiled-coil rod that directly interacts with microtubules in a cell-cycle regulated way. It is an excellent marker for mammalian golgi complex. It is also an antigen in chronic rheumatoid arthritis and in the autoimmune disease Sjoegren syndrome.

  • Stable for at least 1 year after receipt when stored at -20°C.

Purified from HEK 293 cell culture supernatant.

Purity: >95% (SDS-PAGE)

Type:
Antigen: Giantin
Clonality:
Clone: TA10
Conjugation:
Epitope:
Host: Human
Isotype: IgG2 lambda
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR