Anti-PIDD Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25B-0015-C050
102514-672EA 318.46 USD
102514-672
Anti-PIDD Rabbit Polyclonal Antibody
Antibodies
PIDD promotes apoptosis downstream of the tumor suppressor as component of the DNA damage/stress response pathway that connects p53/TP53 to apoptosis. It associates with NEMO/IKBKG and RIP1 and enhances sumoylation and ubiquitination of NEMO/IKBKG which is important for activation of the transcription factor NF-kappa-B. It associates with caspase-2 and RAIDD, and induces activation of caspase-2 which is an important regulator in the apoptotic pathways.

Stable for at least 1 year after receipt when stored at -20°C.

Type:
Antigen: PIDD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR