Anti-TRAM Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25B-0011-R050
102514-666EA 152.31 USD
102514-666
Anti-TRAM Rabbit Polyclonal Antibody
Antibodies
TRAM functions in LPS-TLR4 signaling to regulate the MYD88-independent pathway during the innate immune response to LPS. It is also involved in IL-1-triggered NF-kappaB activation, functioning upstream of IRAK1, IRAK2, TRAF6 and IKBKB. TRAM physically bridges TLR4 and TICAM-1 and functionally transmits LPS-TRL4 signal to TICAM-1. It is expressed in spleen, prostate, testis, uterus, small intestine, colon, peripheral blood leukocytes, heart, placenta, lung, liver, skeletal muscle and pancreas.

Stable for at least 1 year after receipt when stored at -20°C.

Type:
Antigen: TRAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR