Anti-Clusterin Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0099-C100
102514-606EA 346.15 USD
102514-606
Anti-Clusterin Rabbit Polyclonal Antibody
Antibodies
Clusterin is a secretory heterodimeric disulfied-linked glycoprotein that is expressed in virtually all tissues, and found in all human fluids. It is involved in numerous physiological processes important for carcinogenesis and tumor growth, including apoptotic cell death, cell cycle regulation, DNA repair, cell adhesion, tissue remodeling, lipid transportation, membrane recycling, and immune system regulation.

Type:
Antigen: Clusterin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR