Anti-Calreticulin Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0094-C100
102514-596EA 346.15 USD
102514-596
Anti-Calreticulin Rabbit Polyclonal Antibody
Antibodies
Calreticulin is involved in regulation of intracellular Ca2+ homoeostasis and ER Ca2+ capacity. It constitutes together with calnexin and ERp57 the 'calreticulin/calnexin cycle' that is responsible for folding and quality control of newly synthesized glycoproteins. Calreticulin has been implicated to play a role in many biological systems, including functions inside and outside the ER, indicating that the protein is a multi-process molecule.

Stable for at least 6 months after receipt when stored at -20°C.

Type:
Antigen: Calreticulin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR