Anti-RELM-beta Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0063-C100
102514-532EA 221.54 USD
102514-532
Anti-RELM-beta Rabbit Polyclonal Antibody
Antibodies
RELM-beta is a 19kDa disulfide-linked homodimeric protein expressed in the epithelium of the colon and small bowel. RELM-beta has been suggested to play a regulatory role during inflammation and may also act to establish links among adipose tissue, the intestine and the liver. Interestingly, the molecular structure of RELM-beta is highly homologous to that of the adipose-derived cytokine resistin.

Type:
Antigen: RELM-beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR