Anti-ANGPTL3 Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0046-C100
102514-498EA 360 USD
102514-498
Anti-ANGPTL3 Rabbit Polyclonal Antibody
Antibodies
ANGPTL3 regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism.

Type:
Antigen: ANGPTL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR