Anti-Obestatin Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0043-C100
102514-490EA 221.54 USD
102514-490
Anti-Obestatin Rabbit Polyclonal Antibody
Antibodies
Obestatin derives like ghrelin from proghrelin, but exhibits opposite physiological effects. Obestatin decreases food intake and body weight in rodents. Moreover, obestatin also counteracts ghrelin's effect by decelerating gastric emptying and decreasing intestinal contractility. Obestatin binds to orphan G protein-coupled receptor GPR39. Amidation of the C-terminus seems to be important for obestatin's activity.

Type:
Antigen: Obestatin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR