Anti-FABP4 Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0041-C100
102514-484EA 221.54 USD
102514-484
Anti-FABP4 Rabbit Polyclonal Antibody
Antibodies
FABP4 is a fatty acid binding protein in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs regulate the fatty acid uptake, transport, and metabolism. aP2, the mouse form of FABP4, seems to be central to the pathway that links obesity to insulin resistance.

Type:
Antigen: FABP4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR