Anti-ANGPTL6 Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0037-C100
102514-474EA 221.54 USD
102514-474
Anti-ANGPTL6 Rabbit Polyclonal Antibody
Antibodies
ANGPTL6 regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism. The serum levels of ANGPTL6 are significantly increased in obese humans, patients with type 2 diabetes mellitus (T2DM), and in metabolic syndrome.

Type:
Antigen: ANGPTL6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR