Anti-GITR Rat Polyclonal Antibody

Supplier: Adipogen
AG-25A-0017-C100
102514-428EA 346.15 USD
102514-428
Anti-GITR Rat Polyclonal Antibody
Antibodies
GITR is the receptor for TNFSF18. It is involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. Mediates NF-kappaB activation via the TRAF2/NIK pathway.

Type:
Antigen: GITR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR