Anti-RANKL Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0016-C100
102514-426EA 221.54 USD
102514-426
Anti-RANKL Rabbit Polyclonal Antibody
Antibodies
RANKL is an osteoclast differentiation and activation factor. Augments the ability of dendritic cells to stimulate naive T-cell proliferation. Important regulator of interactions between T-cells and dendritic cells and may play a role in the regulation of the T-cell-dependent immune response. Plays an important role in enhanced bone-resorption in humoral hypercalcemia of malignancy.

Type:
Antigen: RANKL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR