Anti-Resistin Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0013-C100
102514-414EA 346.15 USD
102514-414
Anti-Resistin Rabbit Polyclonal Antibody
Antibodies
The adipocytokine resistin which belongs to a family of cysteine-rich C-terminal proteins known as resistin-like molecules (RELM; RELMalpha/FIZZ 1 and RELMbeta/FIZZ 2) of FIZZ (found in inflammatory zone) are thought to be involved in inflammatory processes. Previous studies in mice showed that resistin impairs glucose tolerance and insulin action. In addition, resistin also inhibits adipogenesis in murine 3T3-L1 cells. Therefore resistin has also been proposed as an adipocyte-secreted factor thought to link obesity and T2DM.

Type:
Antigen: Resistin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR