Anti-Adiponectin Rabbit Polyclonal Antibody

Supplier: Adipogen
AG-25A-0003-C100
102514-374EA 346.15 USD
102514-374
Anti-Adiponectin Rabbit Polyclonal Antibody
Antibodies
ACRP30 was identified as a novel adipocyte-specific synthesized and secreted protein with structural resemblance to complement factor C1q. Like adipsin, ACRP30 secretion is induced ~10-fold during adipocyte differentiation. Plasma levels are reduced in obese humans, and low levels are associated with insulin-resistance. Treatment of db/db mice with TZD increased ACRP30 levels.

Type:
Antigen: Adiponectin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR