Anti-NAIP1/2/5 Mouse Monoclonal Antibody [clone: Naipa-1]

Supplier: Adipogen
AG-20B-0045-C100
102514-362EA 540 USD
102514-362
Anti-NAIP1/2/5 Mouse Monoclonal Antibody [clone: Naipa-1]
Antibodies
The inflammasome is a multiprotein complex that mediates the activation of caspase-1, which promotes, amongst others, the secretion of the proinflammatory cytokines interleukin (IL)-1beta and IL-18. Members of the Nod-like receptor (NLR) family are critical components of the inflammasome that link danger-signals to caspase-1 activation. The NLR member IPAF/NLRC4 senses bacterial flagellin and a conserved type III secretion system (TTSS) rod component. A member of the NLR family, NAIP5 directly and specifically interacts with flagellin, triggers a physical NAIP5-NLRC4 association leading to NLRC4 activation. The related NAIP2 functions analogously to NAIP5, serving as a specific inflammasome receptor for TTSS rod proteins such as Salmonella PrgJ and Burkholderia BsaK.

  • Stable for at least 1 year after receipt when stored at -20°C.

Purified from concentrated hybridoma tissue culture supernatant.

Purity: >95% (SDS-PAGE)

Type:
Antigen: NAIP1/2/5
Clonality: Monoclonal
Clone: Naipa-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR