Anti-TACI Rat Monoclonal Antibody (Biotin) [clone: 1A-10]

Supplier: Adipogen
AG-20B-0035B-C100
102514-336EA 484.62 USD
102514-336
Anti-TACI Rat Monoclonal Antibody (Biotin) [clone: 1A-10]
Antibodies
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.

  • Stable for at least 1 year after receipt when stored at -20°C.

Purified from concentrated hybridoma tissue culture supernatant.

Purity: >95% (SDS-PAGE)

Type:
Antigen: TACI
Clonality: Monoclonal
Clone: 1A-10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR