Anti-TRAIL-R2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HS201]

Supplier: Adipogen
AG-20B-0023F-T100
102514-310EA 553.85 USD
102514-310
Anti-TRAIL-R2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HS201]
Antibodies
TRAIL-R2 is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.

  • Purified from concentrated hybridoma tissue culture supernatant.
  • Purity: >95% (SDS-PAGE)

Keep conjugated formats at +4°C.

Type:
Antigen: TRAIL-R2
Clonality: Monoclonal
Clone: HS201
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR