Anti-LAG-3 Mouse Monoclonal Antibody [clone: 17B4]

Supplier: Adipogen
AG-20B-0012-C100
102514-276EA 540 USD
102514-276
Anti-LAG-3 Mouse Monoclonal Antibody [clone: 17B4]
Antibodies
Lymphocyte activation gene 3 (LAG-3; CD223) plays an important role in negatively regulating T cell proliferation, function and homeostasis. It is required for maximal natural and induced regulatory T cell (Treg) function. LAG-3 is closely related to the T cell co-receptor CD4 and binds to MHC class II molecules but with a significantly higher affinity than CD4.

Stable for at least 1 year after receipt when stored at -20°C.

Purity: >95% (SDS-PAGE)

Type:
Antigen: LAG-3
Clonality: Monoclonal
Clone: 17B4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR