Anti-IL-17A Mouse Monoclonal Antibody [clone: 17-142F]

Supplier: Adipogen
AG-20A-0071-C050 AG-20A-0071-C100
102514-168EA 249.23 USD
102514-168 102514-170
Anti-IL-17A Mouse Monoclonal Antibody [clone: 17-142F]
Antibodies
IL-17A exhibits multiple biological activities on a variety of cells including the induction of IL-6 and IL-8 production in fibroblsts, the enhancement of surface expression of ICAM-1 in fibroblast, activation of NF-kappaB and costimulation of T cell proliferation. Induces stromal cells to produce proinflammatory and hematopoietic cytokines.

Type:
Antigen: IL-17A
Clonality: Monoclonal
Clone: 17-142F
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR