Anti-ACE2 Mouse Monoclonal Antibody (Biotin) [clone: AC384]

Supplier: Adipogen
AG-20A-0037B-C050
102514-060EA 346.15 USD
102514-060
Anti-ACE2 Mouse Monoclonal Antibody (Biotin) [clone: AC384]
Antibodies
ACE2 has direct effects on cardiac functions. It is expressed predominantly in vascular endothelial cells of the heart and kidney. ACE2 converts angiotensin I to angiotensin 1-9. ACE2 was also identified as a functional receptor for SARS coronavirus.

Type:
Antigen: ACE2
Clonality: Monoclonal
Clone: AC384
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR