Aranciamycin (from Streptomyces echinatus) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0364-C500 BVT-0364-M001
102513-800EA 180 USD
102513-800 102513-802
Aranciamycin (from Streptomyces echinatus) ≥98% (by HPLC)
Aranciamycin

Aranciamycin is a clostridium histolyticum collagenase inhibitor


  • Antibiotic
  • Anti-tumor compund


Antibiotic. Similar to steffimycin. Active against Gram-positive bacteria.


Yellow to orange solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO

Storage Temperature: Freezer
CAS Number: 72389-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR