Alternariol monomethyl ether ≥98% (by HPLC)

Supplier: 0000027161
Danger

Synonyms: Alternariol-9-methyl ether, 3,7-Dihydroxy-9-methoxy-1-methyl-6H-dibenzo[b,d]pyran-6-one

BVT-0323-C500 BVT-0323-M001
102513-770EA 117.69 USD
102513-770 102513-772
Alternariol monomethyl ether ≥98% (by HPLC)
Alternariol monomethyl ether

Mycotoxin. Antifungal. Phytotoxic. Shown to possess DNA strand-breaking activity. Inhibits the activity of topoisomerase IIalpha (Topo IIalpha) in mammalian cells. Antiviral (Herpes simplex). Hepatitis C NS3-4A protease inhibitor.

Formula: C₁₅H₁₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00058532
CAS Number: 26894-49-5
UN: 3462
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Isolated from Alternaria sp.
Purity >98% (HPLC)
Appearance/Color Off-white to light brown solid.
Solubility Soluble in methanol, dichloromethane or methylene chloride.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR