MM 47755 (from Streptomyces sp. Gö 40/14) ≥98% (by HPLC)

Supplier: 0000027161

BVT-0282-M001
102513-704EA 166.15 USD
102513-704
MM 47755 (from Streptomyces sp. Gö 40/14) ≥98% (by HPLC)
MM 47755

MM 47755 is an anti-bacterial antibiotic.


  • Yellow solid


Antibiotic. Angucyclinone. Anti-bacterial and anti-fungal.


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO, methanol or acetone

Storage Temperature: Freezer
CAS Number: 117620-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR