Concanamycin C (from Streptomyces species) ≥98% (by HPLC), high purity

Supplier: 0000027161

BVT-0254-C025 BVT-0254-C100 BVT-0254-C500
102513-654EA 110.77 USD
102513-654 102513-656 102513-658
Concanamycin C (from Streptomyces species) ≥98% (by HPLC), high purity
Concanamycin C

Concanamycin C is an anti-viral, immunosuppressive antibiotic.


  • Antibiotic
  • Anti-osteoporotic
  • Inhibitor of autophagic degradation


Vacuolar-type H+-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Antifungal and anti-yeast.


White to off-white solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO, methanol, chloroform or acetonitrile; insoluble in water.

Formula: C₄₅H₇₄O₁₃
Storage Temperature: Freezer
CAS Number: 81552-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR