Manumycin A (from Streptomyces parvulus) ≥96% (by HPLC)

Supplier: 0000027161

BVT-0091-M001 BVT-0091-M005
102513-526EA 131.54 USD
102513-526 102513-528
Manumycin A (from Streptomyces parvulus) ≥96% (by HPLC)
Manumycin A

Manumycin A is a potent, selective and competitive cell permeable rasfarnesyltransferase inhibitor (IC50 = 30 nM).


  • Antibiotic
  • Does not affect geranylgeranyltransferase
  • Neutral sphingomyelinase inhibitor


Antibiotic. Does not affect geranylgeranyltransferase (IC50 = 180 µM). Inhibition is competitive with respect to farnesyl pyrophosphate and non-competitive with respect to Ras. Neutral sphingomyelinase inhibitor. Blocks insulin-induced MAP kinase activation in rat cardiac myocytes (19 µM).


Yellow to brown powder


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO or methanol; insoluble in water

Formula: C₃₁H₃₈N₂O₇
Storage Temperature: Freezer
MDL Number: MFCD00920782
CAS Number: 52665-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR