Mutolide (from Sphaeropsidales sp.) ≥98% (by HPLC)

Supplier: 0000027161

BVT-0070-M001
102513-504EA 180 USD
102513-504
Mutolide (from Sphaeropsidales sp.) ≥98% (by HPLC)
Mutolide

Solid


Antibiotic. Antibacterial. Phytotoxic.


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Colour: Off-White

Formula: C₁₄H₂₀O₄
Storage Temperature: Freezer
CAS Number: 277749-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR