Anti-CD4 Monoclonal Antibody [clone: GK1.5]

Supplier: Adipogen
ANC-704-020
102513-398EA 297.69 USD
102513-398
Anti-CD4 Monoclonal Antibody [clone: GK1.5]
Antibodies
CD4 binds to MHC Class II expressed by antigen presenting cells, facilitating T cell receptor signaling.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Rat IgG2bkappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR