Human CD95 (from CHO cells)
Supplier: 0000027161
ANC-506-020
102513-246EA 617.54 USD
102513-246
Human CD95 (from CHO cells)
Proteins and Peptides
Human CD95 (APO-1; Fas) is a type I cell surface glycoprotein that is strongly upregulated on activated T cells, B cells, NK cells and thymocytes. CD95 plays an important role in programmed cell death or apoptosis. Apoptosis appears to be a mechanism for regulating the immune response.