Human CD152
Supplier: 0000027161
ANC-501-050
102513-214EA 1192.15 USD
102513-214
Human CD152
Proteins and Peptides
Human CD152 (CTLA-4) is a cell surface glycoprotein expressed at low levels on activated T cells. CD152 is a high affinity receptor for the costimulatory molecules CD80 (B7-1) and CD86 (B7-2) and appears to function as a negative regulator of T cell activation. This product is a soluble fusion protein combining the extracellular (125aa) domain of human CD152 and murine IgG2a Fc (CTLA-4 Ig).