Anti-CD252 Monoclonal Antibody [clone: ANC10G1]

Supplier: Adipogen
ANC-400-820
102513-198EA 350.31 USD
102513-198
Anti-CD252 Monoclonal Antibody [clone: ANC10G1]
Antibodies
Human CD252 (CD134L; OX40 ligand) is a type II membrane protein with homology to TNF which is expressed on activated B cells and activated endothelial cells. CD252 binds to CD134 (OX40) present on activated T cells, providing a costimulatory signal. Blockade of this interaction in mouse abrogated immunological effects in several models of inflammation and rejection.

  • preservative free

Type: Primary
Antigen: CD252
Clonality: Monoclonal
Clone: ANC10G1
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR