Anti-CD165 Monoclonal Antibody [clone: AD2]

Supplier: Adipogen
ANC-392-820
102513-150EA 297.69 USD
102513-150
Anti-CD165 Monoclonal Antibody [clone: AD2]
Antibodies
Human CD165 is a cell surface molecule present on a subset of peripheral lymphocytes and monocytes and is important for adhesion of thymocytes to thymic epithelial cells.

  • preservative free

Type: Primary
Antigen: CD165
Clonality: Monoclonal
Clone: AD2
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR