Anti-CD147 Monoclonal Antibody [clone: UM-8D6]

Supplier: Adipogen
ANC-376-820
102513-122EA 297.69 USD
102513-122
Anti-CD147 Monoclonal Antibody [clone: UM-8D6]
Antibodies
Human CD147 (EMMPRIN) is a transmembrane glycoprotein expressed more intensely on thymocytes than on mature peripheral blood T cells. Expression of CD147 in bronchial epithelial cells has been correlated with incidence of COPE.

  • preservative free

Type: Primary
Antigen: CD147
Clonality: Monoclonal
Clone: UM-8D6
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR