Anti-CD272 Monoclonal Antibody [clone: ANC5A5]

Supplier: Adipogen
ANC-372-820
102513-112EA 350.31 USD
102513-112
Anti-CD272 Monoclonal Antibody [clone: ANC5A5]
Antibodies
Human CD272 (BTLA; B and T Lymphocyte Attenuator) is a member of the immunoglobulin superfamily and has sequence homology to PD-1 and CTLA-4. It is expressed on T and B lymphocytes and other hemopoetic lineages. Engagement of this molecule by its ligand CD270 (HVEM) can downregulate activated T and B cell responses. CD272 levels on antigen specific CD8+ T cells have been reported to decrease in viral specific, but not melanoma specific activated lines. Polymorphism of the CD272 molecule has been linked to rheumatoid arthritis.

  • preservative free

Type: Primary
Antigen: CD272
Clonality: Monoclonal
Clone: ANC5A5
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR