Anti-CD137 Monoclonal Antibody (R-PE) [clone: 4B4-1]

Supplier: Adipogen
ANC-360-050
102513-090EA 520.62 USD
102513-090
Anti-CD137 Monoclonal Antibody (R-PE) [clone: 4B4-1]
Antibodies
Human CD137 (4-1BB) is expressed on activated T cells within 24-48 hours of activation. CD137 is a type I membrane protein and a member of the TNF receptor superfamily. CD137 appears to be important for T cell proliferation and survival and induces monocyte activation.

Type: Primary
Antigen: CD137
Clonality: Monoclonal
Clone: 4B4-1
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR