Anti-CD152 Monoclonal Antibody [clone: ANC152.2/8H5]

Supplier: Adipogen
ANC-359-020
102513-068EA 350.31 USD
102513-068
Anti-CD152 Monoclonal Antibody [clone: ANC152.2/8H5]
Antibodies
Human CD152 (CTLA-4) is a cell surface glycoprotein expressed at low levels on activated T cells. CD152 is a high affinity receptor for the costimulatory molecules CD80 (B7-1) and CD86 (B7-2) and appears to function as a negative regulator of T cell activation.

Type: Primary
Antigen: CD152
Clonality: Monoclonal
Clone: ANC152.2/8H5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Cat, Cow, Dog, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR