Anti-Perforin Monoclonal Antibody (R-PE) [clone: deltaG9]

Supplier: Adipogen
ANC-358-050
102513-066EA 520.62 USD
102513-066
Anti-Perforin Monoclonal Antibody (R-PE) [clone: deltaG9]
Antibodies
Human perforin (cytolysin) is believed to be one of the major effector molecules used by cytotoxic T cells and NK cells to mediate targeted cell lysis. Perforin expression is constitutive on NK cells, but increases in resting CD8+ cytotoxic cells upon activation.

Type: Primary
Antigen: Perforin
Clonality: Monoclonal
Clone: deltaG9
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2bkappa
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR