Anti-BCL-2 Monoclonal Antibody [clone: Bcl-2/100]

Supplier: Adipogen
ANC-357-020
102513-052EA 350.31 USD
102513-052
Anti-BCL-2 Monoclonal Antibody [clone: Bcl-2/100]
Antibodies
The human bcl-2 gene family plays a central role in lymphoid system cell survival. Continuous expression of bcl-2 protein is critical for preventing apoptosis of mature peripheral B and T cells.

Type: Primary
Antigen: BCL-2
Clonality: Monoclonal
Clone: Bcl-2/100
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR