Anti-CD49F Monoclonal Antibody (Biotin) [clone: BQ16]

Supplier: Adipogen
ANC-356-030
102513-046EA 350.31 USD
102513-046
Anti-CD49F Monoclonal Antibody (Biotin) [clone: BQ16]
Antibodies
Human CD49f molecules are integrin alpha 6 chains that are expressed as heterodimers with CD29 (beta 1 integrin) or CD104 (beta 4 integrin) and function as adhesion receptors. CD49f is expressed on platelets, monocytes, T lymphocytes and thymoctyes.

Type: Primary
Antigen: CD49F
Clonality: Monoclonal
Clone: BQ16
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR