Anti-CD134 Monoclonal Antibody (Biotin) [clone: BerAct35]

Supplier: Adipogen
ANC-355-030
102513-036EA 483.23 USD
102513-036
Anti-CD134 Monoclonal Antibody (Biotin) [clone: BerAct35]
Antibodies
Human CD134 (OX40; ACT35) is an activation-associated antigen which is predominantly expressed on activated CD4 positive cells. CD134 antigen is a member of the TNF receptor family of molecules and may be involved with regulating T cell-dependent B cell proliferation and differentiation.

Type: Primary
Antigen: CD134
Clonality: Monoclonal
Clone: BerAct35
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR