Anti-CD127 Monoclonal Antibody (Biotin) [clone: ANC8F2]

Supplier: Adipogen
ANC-348-030
102513-006EA 483.23 USD
102513-006
Anti-CD127 Monoclonal Antibody (Biotin) [clone: ANC8F2]
Antibodies
CD127 (IL-7Ra) is the specific receptor component for the cytokine interleukin-7 (IL-7). It is found on a wide variety of hematopoietic cell types including B cell precursors and the majority of T cells. Its expression levels are decreased on T cells following activation. CD127 can dimerize with CD132 (IL-2Rgamma) to form a high affinity IL-7 receptor. CD127 engagement is necessary for T cell development in humans.

Type: Primary
Antigen: CD127
Clonality: Monoclonal
Clone: ANC8F2
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR