Anti-CD105 Monoclonal Antibody [clone: SN6]

Supplier: Adipogen
ANC-326-820
102512-968EA 297.69 USD
102512-968
Anti-CD105 Monoclonal Antibody [clone: SN6]
Antibodies
Human CD105 (endoglin) is a homodimeric membrane glycoprotein virtually restricted to endothelial cells and present on some leukemia cells and minor subsets of bone marrow cells. CD105 binds to TGF-beta1 and beta3 with high affinity.

  • preservative free

Type: Primary
Antigen: CD105
Clonality: Monoclonal
Clone: SN6
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR