Anti-CD84 Monoclonal Antibody (Biotin) [clone: 152.1D5]

Supplier: Adipogen
ANC-305-030
102512-890EA 447.23 USD
102512-890
Anti-CD84 Monoclonal Antibody (Biotin) [clone: 152.1D5]
Antibodies
Human CD84 is a cell surface glycoprotein found on mature B lymphocytes, monocytes and a subset of T lymphocytes.

Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: 152.1D5
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR