Anti-CD83 Monoclonal Antibody (Biotin) [clone: HB15e]

Supplier: Adipogen
ANC-304-030
102512-880EA 350.31 USD
102512-880
Anti-CD83 Monoclonal Antibody (Biotin) [clone: HB15e]
Antibodies
Human CD83 is a type 1 cell surface glycoprotein expressed almost solely by dendritic cells that play an important role in antigen presentation.

Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: HB15e
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR