Anti-CD81 Monoclonal Antibody (Biotin) [clone: 1.3.3.22]

Supplier: Adipogen
ANC-302-030
102512-870EA 447.23 USD
102512-870
Anti-CD81 Monoclonal Antibody (Biotin) [clone: 1.3.3.22]
Antibodies
Human CD81 is a member of the tetraspan family of leukocyte cell surface molecules which are involved in signal transduction, cell-cell adhesion and cellular activation. Association of CD81 with integrins appears to be functionally important in the regulation of cell adhesion and activation. Recent work with CD81-null mice indicates CD81 is important for development of humoral immune response.

Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: 1.3.3.22
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR