Anti-CD79B Monoclonal Antibody (R-PE) [clone: SN8]

Supplier: Adipogen
ANC-301-050
102512-864EA 520.62 USD
102512-864
Anti-CD79B Monoclonal Antibody (R-PE) [clone: SN8]
Antibodies
Human CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta) which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg) (3). The mIgM component binds antigen and the Ig alpha/Ig beta heterodimer transduces signals. Analysis of CD79b expression is useful in diagnosis of B cell lymphoproliferative disorders.

Type: Primary
Antigen: CD79B
Clonality: Monoclonal
Clone: SN8
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR