Mouse IgG3 Isotype Control [clone: SIPC 3385] (SIPC 3385) (Biotin)

Supplier: Adipogen
ANC-287-030
102512-822EA 297.69 USD
102512-822
Mouse IgG3 Isotype Control [clone: SIPC 3385] (SIPC 3385) (Biotin)
Antibodies
Immunoglobulin expressed by the mouse plasmacytoma hybridoma SIPC 3385 is an IgG3 isotype of unknown specificity. It is used as a negative control when working with human cells.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: SIPC 3385
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG3kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR