Mouse IgG2b Isotype Control [clone: BPC 4] (BPC 4) (R-PE)

Supplier: Adipogen
ANC-284-050
102512-816EA 350.31 USD
102512-816
Mouse IgG2b Isotype Control [clone: BPC 4] (BPC 4) (R-PE)
Antibodies
Immunoglobulin expressed by the mouse plasmacytoma hybridoma BPC 4 is an IgG2bkappa isotype of unknown specificity. It is used as a negative control when working with human cells.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: BPC 4
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2bkappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR