Mouse IgG2a Isotype Control [clone: RPC 5] (RPC 5) (Biotin)

Supplier: Adipogen
ANC-281-030
102512-800EA 297.69 USD
102512-800
Mouse IgG2a Isotype Control [clone: RPC 5] (RPC 5) (Biotin)
Antibodies
Immunoglobulin expressed by the mouse plasmacytoma hybridoma RPC 5 is an IgG2a isotype of unknown specificity. It is used as a negative control when working with human cells.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: RPC 5
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR